Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyano-7-ethoxycoumarin

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-C0092-M020 CDX-C0092-M100
102989-458EA 254.77 USD InStock
102989-458 102989-460
3-Cyano-7-ethoxycoumarin
3-Cyano-7-ethoxycoumarin
Substrate for determination of cytochrom P-450 dependent oxidases; about 50-100 times more sensitive than conventional substrate ethoxy-resorufin due to higher turn over rate.


Formula: C₁₂H₉NO₃
MDL Number: MFCD00467359
CAS Number: 117620-77-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty