Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diclofenac ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste
Danger

CDX-D0147-G005 CDX-D0147-G025 CDX-D0147-G100
102987-900EA 88.62 USD InStock
102987-900 102987-902 102987-904
Diclofenac ≥98%
Diclofenac
Nonsteroidal anti-inflammatory drug (NSAID) taken or applied to reduce inflammation and as an analgesic reducing pain. The primary mechanism responsible for its anti-inflammatory, antipyretic, and analgesic action is thought to be inhibition of prostaglandin synthesis by inhibition of cyclooxygenase (COX). It also appears to exhibit bacteriostatic activity by inhibiting bacterial DNA synthesis. Can be used as a reference compound.


Formula: C₁₄H₁₁Cl₂NO₂
Storage Temperature: Ambient
CAS Number: 15307-86-5
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Source/Host Synthetic
Purity >98% (HPLC)
Appearance/Color White powder.
Solubility Sligthly soluble in water (2.37 mg/L).

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty