Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Heptafluorobutyryl)imidazole ≥97% (by NMR)

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-H0101-G001 CDX-H0101-G005 CDX-H0101-G025
102990-084EA 70.62 USD InStock
102990-084 102990-086 102990-088
1-(Heptafluorobutyryl)imidazole ≥97% (by NMR)
1-(Heptafluorobutyryl)imidazole
Mild amine-group derivatizing reagent. Non-acidic by-product prevents decomposition and reduces GC column degradation.


Formula: C₇H₃F₇N₂O
Storage Temperature: Freezer
MDL Number: MFCD00014503
CAS Number: 32477-35-3
UN: 1993
ADR: ,
Order Now
Specification Test Results
Source/Host Synthetic
Purity 97% (NMR)
Appearance/Color Yellow liquid.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty