Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isopropylantipyrine

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-P0090-M100 CDX-P0090-G001
102990-814EA 88.62 USD InStock
102990-814 102990-816
4-Isopropylantipyrine
4-Isopropylantipyrine
Analgesic, antipyretic, and anti-inflammatory compound.


Formula: C₁₄H₁₈N₂O
MDL Number: MFCD00053368
CAS Number: 479-92-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty