Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Resorufin-7-O-methyl ether ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-R0012-M001 CDX-R0012-M010 CDX-R0012-M100
102990-902EA 78.92 USD InStock
102990-902 102990-904 102990-906
Resorufin-7-O-methyl ether ≥98%
Resorufin-7-O-methyl ether
Substrate for Cytochrome P-450. Used for various investigations including the differentiatiation of cytochrome P-450 isozymes.


Formula: C₁₃H₉NO₃
MW: 227.219 g/mol
Storage Temperature: Freezer
CAS Number: 5725-89-3
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 98% (TLC)
Appearance/Color Solid.
Solubility Soluble in alcohols, DMSO or DMF.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty