Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Ethoxyresorufin ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-R0013-M001 CDX-R0013-M010 CDX-R0013-M100
102990-908EA 78.92 USD InStock
102990-908 102990-910 102990-912
7-Ethoxyresorufin ≥95%
7-Ethoxyresorufin
Dealkylase substrate for the microfluorimetric analysis of microsomal cytochrome P-450.


Formula: C₁₄H₁₁NO₃
MW: 241.246 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037661
CAS Number: 5725-91-7
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 95% (UV)
Appearance/Color Orange to red solid.
Solubility Soluble in alcohols, DMSO or DMF.

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty