Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Resazurin sodium salt

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-R0051-G005 CDX-R0051-G020
102990-960EA 91.38 USD InStock
102990-960 102990-962
Resazurin sodium salt
Resazurin sodium salt
Fluorescent reagent.


Formula: C₁₂H₆NNaO₄
MW: 251.17 g/mol
MDL Number: MFCD00005036
CAS Number: 62758-13-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty