Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rubratoxin A ≥95% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

AG-CN2-0092-C250 AG-CN2-0092-C500
102987-320EA 456.92 USD InStock
102987-320 102987-322
Rubratoxin A ≥95% (by HPLC)
Rubratoxin A
Mycotoxin. Potent and selective competitive protein phosphatase 2A (PP2A) inhibitor. Does not inhibit PP1, PP2B, PTP1B, ALP and CIP. Antitumor agent. Antimetastatic.


Formula: C₂₆H₃₂O₁₁
Storage Temperature: Freezer
CAS Number: 22467-31-8
Order Now
Specification Test Results
Source/Host Isolated from Penicillium sp.
Purity 95% (HPLC)
Appearance/Color White powder.
Solubility Soluble in methanol, ethanol, acetone or DMSO.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty