Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R,R)-Hymeglusin ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0103-MC05 AG-CN2-0103-M001 AG-CN2-0103-MM25
102987-348EA 200.77 USD InStock
102987-348 102987-350 102987-352
(R,R)-Hymeglusin ≥95%
(R,R)-Hymeglusin
beta-Lactone antibiotic

Source/Host: Isolated from fungal strain Scopulariopsis sp. F-244.
Purity: >95% (HPLC)
Appearance/Color: White to off-white solid.
Solubility: Soluble in methanol, ethanol, DMSO, chloroform or ethyl acetate. Insoluble in water or hexane.

Antibacterial and antifungal. Specific and irreversible inhibitor of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase in vitro and in vivo by covalently modifying the active Cys129 residue of the enzyme. Mevalonate biosynthesis inhibitor.


Formula: C₁₈H₂₈O₅
MW: 324.4 g/mol
Storage Temperature: Freezer
CAS Number: 29066-42-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty