Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amidepsine D ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

AG-CN2-0110-M001 AG-CN2-0110-MM25
102987-382EA 318.46 USD InStock
102987-382 102987-384
Amidepsine D ≥95%
Amidepsine D
Diacylglycerol acyltransferase (DGAT) inhibitor. Excessive accumulation of triacetylglycerol produced by DGAT may cause fatty liver, obesity and hypertriglyceridemia, which may lead to atherosclerosis, diabetes and metabolic disorders.


Formula: C₂₆H₂₄O₁₀
MW: 496.5 g/mol
Storage Temperature: Freezer
CAS Number: 79786-34-8
Order Now
Specification Test Results
Source/Host Isolated from Humcola sp. FO-2942.
Purity 95% (HPLC)
Appearance/Color White to light pink powder.
Solubility Soluble in methanol, benzene or ethyl acetate. Insoluble in water or hexane.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty