Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amauromine ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

AG-CN2-0113-C250 AG-CN2-0113-M001
102987-394EA 69.23 USD InStock
102987-394 102987-396
Amauromine ≥95%
Amauromine
Antibiotic

Source/Host: Isolated from Gymnascella sp.
Purity: >95% (HPLC)
Appearance/Color: Brown solid.
Solubility: Soluble in DMSO, ethanol or methanol.

Hypotensive vasodilator. Calcium channel antagonist. Potent and selective CB1 receptor antagonist.


Storage Temperature: Freezer
CAS Number: 88360-87-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty