Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopenin ≥98% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

AG-CN2-0134-M001 AG-CN2-0134-M005
102988-028EA 180 USD InStock
102988-028 102988-030
Cyclopenin ≥98% (by HPLC)
Cyclopenin
Antimicrobial. Phytotoxic. Specific acetylcholinesterase (AChE) inhibitor.


Formula: C₁₇H₁₄N₂O₃
Storage Temperature: Freezer
CAS Number: 19553-26-5
Order Now
Specification Test Results
Source/Host Isolated from Penicillium sp.
Purity 98% (HPLC)
Appearance/Color White to slightly pink powder.
Solubility Soluble in DMSO, methanol, chloroform or ethyl acetate. Insoluble in water or hexane.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty