Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Andrastin A ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

AG-CN2-0144-C250 AG-CN2-0144-M001
102988-062EA 110.77 USD InStock
102988-062 102988-064
Andrastin A ≥95%
Andrastin A
Mycotoxin. Protein farnesyltransferase (PFTase) inhibitor. Anti-cancer compound. Directly interacts with P-glycoprotein and inhibits the efflux of antitumor agents in drug resistant cells.


Formula: C₂₈H₃₈O₇
MW: 486.6 g/mol
Storage Temperature: Freezer
CAS Number: 174232-42-9
Order Now
Specification Test Results
Source/Host Isolated from Aspergillus fumigatus.
Purity 95% (HPLC)
Appearance/Color Off-white solid.
Solubility Soluble in DMSO, methanol, ethanol, ethyl acetate or chloroform. Insoluble in water or hexane.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty