Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nocardamine ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0150-M001 AG-CN2-0150-M005
102988-086EA 180 USD InStock
102988-086 102988-088
Nocardamine ≥95%
Nocardamine
Antibiotic

Source/Host: Isolated from Streptomyces sp. K04-0144.
Purity: >95% (HPLC)
Appearance/Color: White to off-white solid.
Solubility: Soluble in DMSO, methanol, ethanol or water.

Anti-mycobacterial. Siderophore (iron (Fe) chelating compound). Inducer of morphological changes in insect cells. Antioxidant. Anticancer compound.


Storage Temperature: Freezer
CAS Number: 26605-16-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty