AG-CN2-0157-M001
AG-CN2-0157-M005
102988-110EA
276.92
USD
InStock
102988-110
102988-112
Virustomycin A ≥97%
Virustomycin A
Antibiotic
Source/Host: Isolated from Streptomyces sp. AM-2604.
Purity: >97% (HPLC)
Appearance/Color: Yellow solid.
Solubility: Soluble in ethanol, methanol, acetone or chloroform. Insoluble in water or hexane.
Anti-viral compound. Antifungal (weak). Anti-protozoal (Trypanosoma, Trichomonas).