Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sericic acid ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0402-M005 AG-CN2-0402-M025
102988-150EA 124.62 USD InStock
102988-150 102988-152
Sericic acid ≥95%
Sericic acid
Anti-inflammatory. Antibacterial. Antifungal. Haemolytic agent. Weak phosphodiesterase 4 (PDE4) inhibitor.. Antioxidant.


Formula: C₃₀H₄₈O₆
MW: 504.7 g/mol
Storage Temperature: Freezer
MDL Number: MFCD24849375
CAS Number: 55306-03-1
Order Now
Specification Test Results
Source/Host Isolated from Vochysia divergens.
Purity 95% (NMR)
Appearance/Color White crystalline solid.
Solubility Soluble in DMSO, ethanol or dimethylformamide. Sparingly soluble in water.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty