Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Vitexin ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0425-M005 AG-CN2-0425-M025
102988-234EA 41.54 USD InStock
102988-234 102988-236
Vitexin ≥98%
Vitexin
alpha-Glucosidase inhibitor. Antioxidant. HIF-1 alpha inhibitor. Anti-metastatic. Apoptosis inducer. Tumor suppressor. Adipogenesis inhibitor. Inhibits SIRT6 in vitro. Inhibits polyubiquitin synthesis by the ubiquitin-conjugating enzyme E2-25K. Cardioprotective. Anti-inflammatory. Shows analgesic effect by targeting TRPV1 channel activity. Neuroprotective. Inhibits NMDA receptors.


Formula: C₂₁H₂₀O₁₀
MW: 432.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00017456
CAS Number: 3681-93-4
Order Now
Specification Test Results
Source/Host Isolated from Crataegus pinnatifida.
Purity 98% (HPLC)
Appearance/Color Yellow crystalline powder.
Solubility Soluble in DMSO or dimethylformamide.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty