Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Herbimycin A ≥99% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0429-C100 AG-CN2-0429-M001
102988-250EA 207.69 USD InStock
102988-250 102988-252
Herbimycin A ≥99% (by HPLC)
Herbimycin A
Benzoquinone ansamycin antibiotic

Source/Host: Isolated from Streptomyces sp.
Purity: >99% (HPLC)
Appearance/Color: Yellow solid.
Solubility: Soluble in ethanol, methanol DMSO or dimethylformamide. Poorly soluble in water.

Herbicidal compound. Potent, selective, irreversible and cell permeable protein tyrosine kinase inhibitor. Inhibitor of v-Src, Yes, Fps, Ros, Bcr-Abl and ErbB oncogene products. Antitumor compound. Antiangiogenic. Inhibits NF-kappaB activation and phosphorylation of phospholipase C-gamma1. Inhibitor of heat shock protein 90 (Hsp90). Binds Hsp90 and destabilizes client proteins leading to their ubiquitination and proteasomal degradation. Increases the sensitivity of certain cancer cells to chemotherapeutic agents. Neuroprotective. Antiparasitic, antischistosomal agent. Antiviral. Increase microtubules sensitivity to cold in plants.


Formula: C₃₀H₄₂N₂O₉
Storage Temperature: Freezer
MDL Number: MFCD00151702
CAS Number: 70563-58-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty