Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclosporin C ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste
Danger

AG-CN2-0443-M001 AG-CN2-0443-M005
102988-282EA 69.23 USD InStock
102988-282 102988-284
Cyclosporin C ≥95%
Cyclosporin C
Immunosuppressant. Calcineurin inhibitor with lower activity than Cyclosporin A (AG-CN2-0079). Antifungal. Antitumor.


Formula: C₆₂H₁₁₁N₁₁O₁₃
MW: 1218.6 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06410945
CAS Number: 59787-61-0
Order Now
Specification Test Results
Source/Host Isolated from Fusarium solani.
Purity 95% (HPLC)
Appearance/Color White solid.
Solubility Soluble in 100% ethanol, acetone or ethyl acetate; insoluble in water.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty