Z-Leu-Leu-Phe-CHO [MG-110]
Supplier: Van Der Voort Lucht en Milieu Amste
AG-CP3-0021-M001 AG-CP3-0021-M005
102988-492EA 83.08 USD InStock
102988-492 102988-494
Z-Leu-Leu-Phe-CHO [MG-110]
Proteins and Peptides
Reversible and cell permeable proteasome inhibitor. Specifically inhibits the chymotrypsin-like activity of the proteasome. Calpain and cathepsin inhibitor. Does not inhibit the peptidyl-glutamyl peptide hydrolyzing activity of MPC.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty