Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

IWR-1-endo ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CR1-3581-M005 AG-CR1-3581-M025
102988-638EA 124.62 USD InStock
102988-638 102988-640
IWR-1-endo ≥98%
IWR-1-endo
Potent and reversible cell permeable Wnt pathway signaling inhibitor. Inhibits Wnt-induced accumulation of beta-catenin, leading to proteasomal degradation of this protein through a destruction complex which consists of Apc, Axin2, CK1 and GSK-3beta. Stabilizes the destruction complex, increasing the level of Axin2 protein without changing the levels of Apc or GSK-3beta. Tankyrase-1 (TNKS1/PARP5a) and Tankyrase-2 (TNKS2/PARP5b) inhibitor (in vitro auto-PARsylation assay).


Formula: C₂₅H₁₉N₃O₃
MW: 409.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18086875
CAS Number: 1127442-82-3
Order Now
Specification Test Results
Purity 98%
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or DMF.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty