Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tofacitinib ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste
Danger

AG-CR1-3625-M005 AG-CR1-3625-M025
102988-814EA 83.08 USD InStock
102988-814 102988-816
Tofacitinib ≥98%
Tofacitinib
Potent cell permeable JAK3 inhibitor (IC50 = 1nM). Shows also JAK-1 inhibitory activity. Immunosuppressive and anti-inflammatory agent. Inhibits signaling through heterodimeric receptors associated with JAK3, JAK1, or both of them, with functional selectivity over JAK2-paired receptors. Blocks downstream STAT signaling, resulting in potent inhibition of several cytokines, including interleukins 2, 4, 7, 9, 15 and 21, which are integral to lymphocyte activation, function and proliferation. Investigated for several autoimmune disorders including, rheumatoid arthritis and psoriasis. Potent and selective inhibitor of HIV-1 replication and virus reactivation in vitro.


Formula: C₁₆H₂₀N₆O
Storage Temperature: Freezer
MDL Number: MFCD11035919
CAS Number: 477600-75-2
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Purity 98%
Appearance/Color Off-white to pale yellow solid.
Solubility Soluble in DMSO or ethanol.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty