Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Averantin ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

BVT-0169-M001 BVT-0169-M005
102988-898EA 110.77 USD InStock
102988-898 102988-900
Averantin ≥98%
Averantin
Averantin is a cytotoxic against human solid tumor cell lines.

  • Mycotoxin
  • Cytotoxic
  • Intermediate to Aflatoxin B1

Intermediate of the biosynthetic pathway to Aflatoxin B1. Cytotoxic against human solid tumor cell lines.

Orange powder

Soluble in acetone, methanol or DMSO


Formula: C₂₀H₂₀O₇
MW: 372.4 g/mol
Storage Temperature: Freezer
CAS Number: 5803-62-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty