Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Verruculogen ≥98% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste
Danger

BVT-0443-M001 BVT-0443-M005
102989-036EA 152.31 USD InStock
102989-036 102989-038
Verruculogen ≥98% (by HPLC)
Verruculogen
Fumitremorgin-type alkaloid. Tremorgenic neurotoxic mycotoxin. Selective blocker of KCa1.1 (high-conductance Ca2+-activated K+; BK; maxi-K) channels. Decreases GABA levels in CNS. Increases spontaneous glutamate and aspartate release. At toxic doses, increases the number and decreases the affinity of DHP receptors in rat cortex. Cell cycle inhibitor at the M phase. Inhibits calmodulin and PKC activity. Has apoptotic effects on human T lymphocytes. Stimulates the growth of seedling roots of buckwheat.


Formula: C₂₇H₃₃N₃O₇
Storage Temperature: Freezer
CAS Number: 12771-72-1
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Source/Host Isolated from Aspergillus fumigatus.
Purity 98% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO, 100% ethanol, methanol or dichloromethane.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty