Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Anthracenylsulfonyl chloride ≥90% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste
Danger

CDX-A0062-M100 CDX-A0062-G001
102989-170EA 254.77 USD InStock
102989-170 102989-172
2-Anthracenylsulfonyl chloride ≥90% (by HPLC)
2-Anthracenylsulfonyl chloride
Derivatizing reagent with strong fluorescence.


Formula: C₁₄H₉ClO₂S
Storage Temperature: Freezer
CAS Number: 17407-98-6
UN: 3261
ADR: 8,III
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 90% (HPLC)
Appearance/Color Yellow powder.
Solubility Soluble in acetonitrile.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty