Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino TAMRA ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-A0156-M010 CDX-A0156-M100
102989-206EA 275.54 USD InStock
102989-206 102989-208
5-Amino TAMRA ≥95%
5-Amino TAMRA
A synthetic intermediate.


Formula: C₂₄H₂₃N₃O₃
Storage Temperature: Refrigerator
CAS Number: 167095-10-5
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 95%
Appearance/Color Purple powder.
Solubility Soluble in DMSO or methanol.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty