Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Azimsulfuron ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-A0203-M100 CDX-A0203-G001
102989-266EA 221.54 USD InStock
102989-266 102989-268
Azimsulfuron ≥98%
Azimsulfuron
Reference standard.


Formula: C₁₃H₁₆N₁₀O₅S
MW: 424.4 g/mol
Storage Temperature: Refrigerator
CAS Number: 120162-55-2
Order Now
Specification Test Results
Source/Host Synthetic.
Purity >98% (HPLC)
Appearance/Color Solid.
Solubility Soluble in acetonitrile.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty