Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bismerthiazol ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-B0246-G001 CDX-B0246-G010
102989-430EA 213.23 USD InStock
102989-430 102989-432
Bismerthiazol ≥98%
Bismerthiazol
Reference standard.


Formula: C₅H₆N₆S₄
MW: 278.4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01681864
CAS Number: 79319-85-0
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty