Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Biotinyl tyramide ≥97%

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-B0270-M100 CDX-B0270-M500
102987-498EA 228.46 USD InStock
102987-498 102987-500
Biotinyl tyramide ≥97%
Biotinyl tyramide


Formula: C₁₈H₂₅N₃O₃S
MW: 363.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD27953026
CAS Number: 41994-02-9
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color White to light pink solid.
Solubility Soluble in DMSO or acetonitrile.

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty