Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-ROX ≥90% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-C0084-M025 CDX-C0084-M125
102989-450EA 333.69 USD InStock
102989-450 102989-452
6-ROX ≥90% (by HPLC)
6-ROX
This pure-6 isomer is a derivative of carboxy-X-rhodamine, widely used for oligonucleotide labeling and automated DNA sequencing applications.


Formula: C₃₃H₃₀N₂O₅
Storage Temperature: Refrigerator
CAS Number: 194785-18-7
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 90% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO or methanol.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty