Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-TET dipivaloate ≥98% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-C0186-M500 CDX-C0186-GG25
102989-536EA 343.38 USD InStock
102989-536 102989-538
6-TET dipivaloate ≥98% (by HPLC)
6-TET dipivaloate
6-TET is an amino-conjugatable fluorescent probe that is widely used in nucleic acid sequencing and related research. It can also be used to label peptides and oligonucleotides.


Formula: C₃₁H₂₄Cl₄O₉
Storage Temperature: Refrigerator
CAS Number: 314734-87-7
Order Now
Specification Test Results
Source/Host Synthetic.
Purity >98% (HPLC); 1% Iso 5
Appearance/Color Solid.
Solubility Soluble in DMSO or DMF.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty