Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(+)-(8,8-Dichlorocamphorylsulfonyl)oxaziridine ≥97%

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-D0031-G005 CDX-D0031-G025
102987-672EA 263.08 USD InStock
102987-672 102987-674
(+)-(8,8-Dichlorocamphorylsulfonyl)oxaziridine ≥97%
(+)-(8,8-Dichlorocamphorylsulfonyl)oxaziridine
Chiral oxidizing agent. Used for asymmetric synthesis as chiral catalyst. Hydroxylation reagent.


Formula: C₁₀H₁₃Cl₂NO₃S
MW: 298.19 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00243015
CAS Number: 127184-05-8
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 97% (Titration)
Appearance/Color Solid.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty