Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Diethylaminocoumarin-3,4-dicarboxylic acid ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-D0039-M100 CDX-D0039-G001
102987-698EA 278.31 USD InStock
102987-698 102987-700
7-Diethylaminocoumarin-3,4-dicarboxylic acid ≥95%
7-Diethylaminocoumarin-3,4-dicarboxylic acid
Proteomic research material.


Formula: C₁₅H₁₅NO₆
MW: 305.28 g/mol
Storage Temperature: Refrigerator
CAS Number: 75240-77-6
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty