Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

FAMC ≥95% (by NMR)

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-D0099-M010 CDX-D0099-M050
102987-796EA 146.77 USD InStock
102987-796 102987-798
FAMC ≥95% (by NMR)
FAMC
Polarity fluorescent probe.


Formula: C₁₃H₈F₂N₄O₃
Storage Temperature: Freezer
CAS Number: 127192-67-0
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 95% (NMR)
Appearance/Color Solid.
Solubility Soluble in DMSO or chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty