Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethylvinphos ≥97%

Supplier: Van Der Voort Lucht en Milieu Amste
Danger

CDX-D0316-M050 CDX-D0316-M250
102989-762EA 193.85 USD InStock
102989-762 102989-764
Dimethylvinphos ≥97%
Dimethylvinphos
Organophosphorus insecticide. Compound can be used as analytical reference material.


Formula: C₁₀H₁₀Cl₃O₄P
MW: 331.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00272599
CAS Number: 2274-67-1
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color Crystalline light red.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty