Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Dehydrocholesterol ≥95% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-D0331-G010 CDX-D0331-G025
102989-778EA 113.54 USD InStock
102989-778 102989-780
7-Dehydrocholesterol ≥95% (by HPLC)
7-Dehydrocholesterol
Precursor of cholesterol and vitamin D3. It is reduced to cholesterol by the enzyme 3beta-hydroxysterol-Delta7-reductase (DHCR7) in the last step of cholesterol biosynthesis. Mutations in the gene encoding DHCR7 lead to increased levels of 7-DHC and the neurodevelopmental syndrome Smith-Lemli-Opitz syndrome (SLOS). 7-DHC is used as a biomarker for SLOS screenings. Inhibits human fibroblast sphingomyelinase. Shown to have cytotoxic effects on melanoma cells. In skin, photolysis of 7-DHC by ultraviolet light produces vitamin D3. UV/Vis: lambdamax 271, 282, 293nm.


Formula: C₂₇H₄₄O
Storage Temperature: Freezer
MDL Number: MFCD00003624
CAS Number: 434-16-2
Order Now
Specification Test Results
Source/Host Synthetic
Purity 95% (HPLC)
Appearance/Color White powder.
Solubility Soluble in ethanol, chloroform or ethyl acetate. Slightly soluble in DMSO. Insoluble in water.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty