Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Ethoxy-4-trifluoromethylcoumarin ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-E0003-M050 CDX-E0003-M250
102989-866EA 213.23 USD InStock
102989-866 102989-868
7-Ethoxy-4-trifluoromethylcoumarin ≥98%
7-Ethoxy-4-trifluoromethylcoumarin
Substrate for monitoring the activity of cytochrome P450 in diverse cells and research issues, including in vitro cytotxity testing, regulation and drug metabolization.


Formula: C₁₂H₉F₃O₃
MW: 258.19 g/mol
Storage Temperature: Refrigerator
CAS Number: 115453-82-2
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 98% (TLC)
Appearance/Color White powder.
Solubility Soluble in methanol, DMSO or DMF.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty