Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fluo-3 ≥70% (by NMR)

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-F0056-M001 CDX-F0056-M005
102989-974EA 279.69 USD InStock
102989-974 102989-976
Fluo-3 ≥70% (by NMR)
Fluo-3
Fluorescent indicator for calcium used in flow cytometry. Fluorescence intensity increases about 40-fold after calcium binding.


Formula: C₃₆H₃₀Cl₂N₂O₁₃
Storage Temperature: Freezer
CAS Number: 123632-39-3
Order Now
Specification Test Results
Source/Host Synthetic.
Purity ~70% (NMR)
Appearance/Color Crystalline solid.
Solubility Soluble in water.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty