Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl fumaroyl chloride ≥97%

Supplier: Van Der Voort Lucht en Milieu Amste
Danger

CDX-F0210-G005 CDX-F0210-G050
102990-018EA 368.31 USD InStock
102990-018 102990-020
Ethyl fumaroyl chloride ≥97%
Ethyl fumaroyl chloride
Reagent for synthesis.


Formula: C₆H₇ClO₃
MW: 162.57 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03002066
CAS Number: 26367-48-6
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 97% (GC)
Appearance/Color Solid.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty