Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxyhexanoyl-L-homoserine lactone ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-H0083-M005 CDX-H0083-M025
102990-278EA 123.23 USD InStock
102990-278 102990-280
3-Hydroxyhexanoyl-L-homoserine lactone ≥98%
3-Hydroxyhexanoyl-L-homoserine lactone
3-Hydroxy-hexanoyl-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.


Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
Storage Temperature: Freezer
CAS Number: 192883-16-2
Order Now
Specification Test Results
Source/Host Synthetic
Purity 98% (NMR)
Appearance/Color White to pink powder.
Solubility Soluble in ethanol, methanol, DMF or DMSO. Poorly soluble in water.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty