Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Halofenozide ≥98% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-H0097-M050 CDX-H0097-M250
102990-074EA 245.08 USD InStock
102990-074 102990-076
Halofenozide ≥98% (by HPLC)
Halofenozide
Dibenzoylhydrazine based nonsteroidal insecticide. Ecdysone agonist that mimics the biological function of the natural insect molting hormone 20-hydroxyecdysone, causing premature molting and interfering with the normal growth and development in insects. Ecdysteroid agonist are also used as ligands for gene expression and gene therapy research. Compound can be used as analytical reference material.


Formula: C₁₈H₁₉ClN₂O₂
Storage Temperature: Freezer
MDL Number: MFCD03158909
CAS Number: 112226-61-6
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White to off-white powder.
Solubility Soluble in water.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty