Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Hydroxy-1,2-benziodoxol-3-one 1-oxide pyridinium complex ≥97% (by NMR)

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-H0104-G001 CDX-H0104-G005
102990-098EA 70.62 USD InStock
102990-098 102990-100
1-Hydroxy-1,2-benziodoxol-3-one 1-oxide pyridinium complex ≥97% (by NMR)
1-Hydroxy-1,2-benziodoxol-3-one-1-oxide pyridinium complex
Stable pyridinium salt of o-iodoxybenzoic acid (PIBX) is a convenient substitute of o-iodoxybenzoic acid (IBX) as an oxidant. PIBX is safer, has neutral properties behaves as an equivalent to IBX in the oxidation of alcohols to ketones or aldehydes in polar solvents and provides higher oxidation rate in THF due to better solubility.


Formula: C₁₂H₁₀INO₄
Storage Temperature: Freezer
CAS Number: 1380548-11-7
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 97% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in toluene.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty