Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7β-Hydroxycholesterol ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-H0116-M005 CDX-H0116-M010 CDX-H0116-M100
102990-118EA 210.46 USD InStock
102990-118 102990-120 102990-122
7β-Hydroxycholesterol ≥98%
7beta-Hydroxycholesterol
Oxysterole. Cholesterol oxidation metabolite. Induces cell death/apoptosis and shows antiproliferative activity against cancer cells, involving mitochondrial perturbation, oxidative stress and lysosomal destabilization. Its membrane organizing properties may have implications in Alzheimer’s disease. Increased levels correlate with risk of cardiovascular diseases including atherosclerosis. Shown to have proinflammatory activity.


Formula: C₂₇H₄₆O₂
MW: 402.65 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058406
CAS Number: 566-27-8
Order Now
Specification Test Results
Source/Host Synthetic
Purity 98% (HPLC)
Appearance/Color White to white-yellow powder.
Solubility Soluble in ethanol, chloroform or methanol.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty