Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

IPB ≥90%

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-I0004-M250 CDX-I0004-G001
102990-144EA 228.46 USD InStock
102990-144 102990-146
IPB ≥90%
IPB
4-[3-(4-iodophenyl)-2-(2,4-dinitrophenyl)-2H-5-tetrazolio]-1,3-benzenedisulfonate sodium salt (IPB) have found application in clinical assays and cell proliferation assays that are based on a spectrophotometric measurement of dehydrogenase activities of cells. Disulfonated tetrazolium salt IPB, that produces a highly water-soluble formazan dye that proved to be extremely useful as a cell-viability indicator. IPB is reduced by reduced nicotinamide adenine dinucleotide (NADH) to formazan in the presence of a redox mediator such as 1-methoxy PMS, which provides the basis to specrophotometric assays of NADH.


Formula: C₁₉H₁₀IN₆NaO₁₀S₂
MW: 696.34 g/mol
Storage Temperature: Freezer
CAS Number: 161617-45-4
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 90% (HPLC)
Appearance/Color Pale yellow powder.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty