Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(3-Isothiocyanatobenzyl)-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium bromide ≥99% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-I0022-M050 CDX-I0022-M250
102990-168EA 193.85 USD InStock
102990-168 102990-170
N-(3-Isothiocyanatobenzyl)-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium bromide ≥99% (by HPLC)
N-(3-Isothiocyanatobenzyl)-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium bromide
New class of highly photostable, water soluble fluorescent labels with large Stoke's shift, high QY in aqueous media and pH tolerance.


Formula: C₂₃H₁₈BrN₃O₂S
Storage Temperature: Freezer
CAS Number: 155862-89-8
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 99% (HPLC)
Appearance/Color Solid.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty