Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lactofen ≥99%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-L0013-M100 CDX-L0013-M500 CDX-L0013-G005
102990-330EA 131.54 USD InStock
102990-330 102990-332 102990-334
Lactofen ≥99%
Lactofen
Analytical standard for chromatography. Environmental standard. Herbicide.


Formula: C₁₉H₁₅ClF₃NO₇
MW: 461.77 g/mol
Storage Temperature: Refrigerator
CAS Number: 77501-63-4
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 99% (HPLC)
Appearance/Color Amber-colored solid or liquid.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty