Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-2-(2-naphthyl)ethanenitrile ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-M0072-M050 CDX-M0072-M250
102990-430EA 202.15 USD InStock
102990-430 102990-432
2-Methoxy-2-(2-naphthyl)ethanenitrile ≥95%
2-Methoxy-2-(2-naphthyl)ethanenitrile
Fluorescent dye.


Formula: C₁₃H₁₁NO
MW: 197.23 g/mol
Storage Temperature: Freezer
CAS Number: 118736-08-6
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in acetonitrile.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty