Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-O-Methylfluorescein ≥97%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-M0098-M005 CDX-M0098-M010 CDX-M0098-M050
102990-492EA 148.15 USD InStock
102990-492 102990-494 102990-496
3'-O-Methylfluorescein ≥97%
3'-O-Methylfluorescein
Used in fluorimetric studies. Fluorescent probe. Selective fluorometric substrate for CYP2C19 and CYP1A1. Reaction product of the substrate 3-O-methylfluorescein phosphate (OMFP) is hydrolyzed by phosphatases to yield O-methylfluorescein (OMF) that is detected at a 485nm excitation/525nm emission.


Formula: C₂₁H₁₄O₅
MW: 346.33 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00055945
CAS Number: 70672-05-8
Order Now
Specification Test Results
Source/Host Synthetic
Purity 97% (HPLC)
Appearance/Color Light red to brown powder.
Solubility Soluble in DMSO.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty