Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Methylumbelliferyl)maleinimide ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-M0117-M050 CDX-M0117-M250
102990-502EA 228.46 USD InStock
102990-502 102990-504
N-(4-Methylumbelliferyl)maleinimide ≥95%
N-(4-Methylumbelliferyl)maleinimide
Fluorescent reagent with high selectivity.


Formula: C₁₄H₉NO₄
MW: 255.23 g/mol
Storage Temperature: Refrigerator
CAS Number: 211565-47-8
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Light yellow powder.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty