Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

MCPA-thioethyl ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-M0136-M050 CDX-M0136-M500
102990-542EA 139.85 USD InStock
102990-542 102990-544
MCPA-thioethyl ≥98%
MCPA-thioethyl
Phenoxy herbicide. Synthetic auxin. Compound can be used as analytical reference material.


Formula: C₁₁H₁₃ClO₂S
MW: 244.7 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01671920
CAS Number: 25319-90-8
UN: 3077
ADR: 9 III
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White to light yellow/beige powder.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty